missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Sialyltransferase Inhibitor, 3Fax-Peracetyl Neu5Ac

Catalog No. 56622410MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56622410MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56622410MG Supplier MilliporeSigma™ Supplier No. 56622410MG

The Sialyltransferase Inhibitor, 3Fax-Peracetyl Neu5Ac controls the biological activity of Sialyltransferase.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach

Specifications

Color White
Description 98% by NMR
Target Proteins Sialyltransferase
Formula Weight 551.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg