missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ SATA

Catalog No. 573100100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
573100100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 573100100MG Supplier MilliporeSigma™ Supplier No. 573100100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reacts with primary amines to add protected sulfhydryl groups.

EMD Millipore Calbiochem™ SATA is used as agent for introducing thiol groups.

  • Conjugates free sulfhydryl group to maleimide-modified proteins, when treated with hydroxylamine
  • Assay: ≥98% by HPLC
  • CAS: 76931-93-6
  • Molecular Formula: C8H9NO5S
  • Packaging: Glass Bottle
  • Color: White

Recommended Storage

−20°C

Specifications

Quantity 100 mg
Product Type SATA, heterobifunctional
Synonym N-Succinimidyl-S-acetylthioacetate
Content And Storage -20°C