missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ RS 127445 Chlorhydrate

Numéro de catalogue. 5081470001
Modifier l'affichage
Click to view available options
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
5081470001 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 5081470001 Fournisseur MilliporeSigma™ Code fournisseur 5.08147.0001
Désactive avec 5% d’eau de Javel

Spécifications

Color Blanc cassé
Description 98% by HPLC
Target Proteins 5-HT2B
Molecular Formula C17H16FN3·HCl
Solubility DMSO ou EtOH
Content And Storage Protège de la lumière
Form Solide
Quantity 10 mg