missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Ro-20-1724

Catalog No. 557502100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
557502100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 557502100MG Supplier MilliporeSigma™ Supplier No. 557502100MG

A cell-permeable, selective inhibitor of cAMP-specific phosphodiesterase (PDE IV; IC50 = 2 μM).

Cell Permeable

Specifications

Color White
Description 98% by TLC
Target Proteins PDE IV
CAS 29925-17-5
Molecular Formula C15H22N2O3
Formula Weight 278.4g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 100 mg