missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ RIPK3 Inhibitor, GSK'872

Catalog No. 5303890001
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5303890001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5303890001 Supplier MilliporeSigma™ Supplier No. 5.30389.0001

A cell-permeable, selective inhibitor of RIP3-selective kinase with >1,000-fold selectivity over a vast majority of more than 300 other kinases.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach

Specifications

Color Yellow
Description 98% by HPLC
Target Proteins RIP3 Kinase
Molecular Formula C19H17N3O2S2
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg