missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Ribonuclease Inhibitor, Human, Recombinant, E. coli

Catalog No. 5568812500U
Change view
Click to view available options
Quantity:
2500 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5568812500U 2500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5568812500U Supplier MilliporeSigma™ Supplier No. 5568812500U

The Ribonuclease Inhibitor, Human, Recombinant, E. coli controls the biological activity of Ribonuclease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Contaminants: Endonuclease and RNase: none detected

Specifications

Concentration 20,000 units/mL
Formulation In 50 mM KCl, 20 mM HEPES, 8 mM DTT, 50% glycerol.
Target Proteins inactivates RNase
Formula Weight 49847g/mol
Form Liquid
Quantity 2500 U