missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Rac Inhibitor III, EHop-016
Description
Packaged Under Inert Gas
Specifications
Specifications
| Color | Pale Yellow |
| Description | 96% by HPLC |
| Product Type | Inhibitor |
| CAS | 1380432-32-5 |
| Molecular Formula | C25H30N6O |
| Formula Weight | 430.6g/mol |
| Solubility | DMSO |
| Content And Storage | Protect from light |
| Form | Powder |
| Quantity | 10 mg |