missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Rabbit γ-Globulin, Serum

Catalog No. 3459915GM
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3459915GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3459915GM Supplier MilliporeSigma™ Supplier No. 3459915GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Native γ-globulin from rabbit serum.

  • Form: Lyophilized
  • Purity: ≥95% by electrophoresis

Specifications

Quantity 5 g
Source Rabbit
Content And Storage Ok to freeze.
Form Lyophilized
Purity or Quality Grade ≥95% by electrophoresis
Host Species Rabbit
Product Type Rabbit γ-Globulin, Serum