missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Proteinase K Inhibitor

Catalog No. 53947010MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53947010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53947010MG Supplier MilliporeSigma™ Supplier No. 53947010MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Proteinase K Inhibitor is a tetrapeptidyl chloromethyl ketone compound that acts as an active-site-targeting irreversible inhibitor against proteinase K.

  • Packaged Under Inert Gas
  • Peptide Sequence: MeOSuc-Ala-Ala-Pro-Phe-CH2Cl

Specifications

Description 97% by HPLC
Molecular Formula C26H35ClN4O7
Formula Weight 551g/mol
Solubility DMSO
Content And Storage Protect from Light, Protect from Moisture
Form Lyophilized
Quantity 10 mg