Learn More
MilliporeSigma™ Calbiochem™ Protease Inhibitor Cocktail Set III, EDTA-Free
Catalog No. 5391341SET
Change view
Click to view available options
Quantity:
1 Set
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5391341SET 1 Set
5391341ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5391341SET Supplier MilliporeSigma™ Supplier No. 5391341SET
Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.
A cocktail of six protease inhibitors with broad specificity for the inhibition of aspartic, cysteine, and serine proteases as well as aminopeptidases. This cocktail is recommended for use with mammalian cell and tissue extracts, but is also suitable for bacterial extracts. Supplied with a data sheet.
Specifications
Target Proteins Aspartic, cysteine, and serine proteases as well as aminopeptidases
Quantity 1 Set