missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Prodigiosin Serratia marcescens

Catalog No. 529685200UG
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
529685200UG 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 529685200UG Supplier MilliporeSigma™ Supplier No. 529685200UG

A cell-permeable tripyrrole alkaloid compound that displays immunosuppressive and anti-tumor properties irrespective of p53 status and multidrug resistance.

Packaged Under Inert Gas

Specifications

Color Dark Red
Description 95% by HPLC
CAS 82-89-3
Molecular Formula C20H25N3O
Formula Weight 323.4g/mol
Solubility DMSO, DMF, EtOH, MeOH
Content And Storage Protect from light
Form Solid
Quantity 200 μg