missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ PLD Inhibitor, FIPI
Description
Packaged Under Inert Gas
Specifications
Specifications
| Color | Off-white |
| Description | 99% by HPLC |
| Product Type | Inhibitor |
| Molecular Formula | C23H24FN5O2 |
| Formula Weight | 421.19g/mol |
| Solubility | DMSO |
| Content And Storage | Protect from light |
| Form | Powder |
| Quantity | 5 mg |