missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ PKCζ Pseudosubstrate Inhibitor, Myristoylated

Catalog No. 539624500UG
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
539624500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 539624500UG Supplier MilliporeSigma™ Supplier No. 539624500UG

PKCζ Pseudosubstrate Inhibitor is a myristoylated, cell-permeable, reversible, substrate competitive inhibitor of protein kinase Cζ.

  • Peptide Sequence: Myr-Ser-Ile-Tyr-Arg-Arg-Gly-Ala-Arg-Arg-Trp-Arg-Lys-Leu-OH
  • Cell Permeable

Specifications

Formulation Supplied as a trifluoroacetate salt.
Description 97% by HPLC
Target Proteins insulin-stimulated transport in intact adinocytes
Molecular Formula C90H154N30O17
Formula Weight 1928.4g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 500 μg