missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ PKA Inhibitor 14-22 Amide, Cell-Permeable, Myristoylated

Catalog No. 476485500UG
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
476485500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 476485500UG Supplier MilliporeSigma™ Supplier No. 476485500UG

PKA Inhibitor 14-22 Amide is myristoylated at the N-terminus that enhances its cell-permeability. The non-myristoylated version is shown to be a specific inhibitor of PKA (Ki = 36 nM).

  • Peptide Sequence: Myr-N-Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-NH2
  • Cell Permeable

Specifications

Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins cAMP-dependent protein kinase
Molecular Formula C53H100N20O12
Formula Weight 1209.5g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 500 μg