missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Phosphatase Inhibitor Cocktail Set II

Catalog No. 5246251SET
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5246251SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5246251SET Supplier MilliporeSigma™ Supplier No. 5246251SET
Only null left

A cocktail of five phosphatase inhibitors for the inhibition of acid and alkaline phosphatases as well as protein tyrosine phosphatases (PTPs). Suitable for use with cell lysates and tissue extracts.

A cocktail of five phosphatase inhibitors for the inhibition of acid and alkaline phosphatases as well as protein tyrosine phosphatases (PTPs). Suitable for use with cell lysates and tissue extracts, including samples containing detergents. Provided as a set of five 1mL vials containing the following components: Imidazole (200 mM), Sodium Fluoride (100 mM), Sodium Molybdate (115 mM), Sodium Orthovanadate (100 mM), and Sodium Tartrate Dihydrate (400 mM).

Specifications

Formulation In H2O.
Form Liquid
Quantity 1 Set