missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ o-Phenanthroline
Description
Packaged Under Inert Gas
Specifications
Specifications
| Color | Off-white |
| Description | 99% by titration |
| Product Type | Inhibitor |
| Target Proteins | Metalloprotease inhibitor |
| CAS | 5144-89-8 |
| Molecular Formula | C12H8N2·H2O |
| Formula Weight | 198.2g/mol |
| Solubility | DMSO, EtOH, or H2O |
| Content And Storage | Protect from light |
| Form | Solid |
| Show More |