missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ NS8593

Catalog No. 49203110MG
Change view
Click to view available options
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
49203110MG 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 49203110MG Supplier MilliporeSigma™ Supplier No. 49203110MG

An aminobenzimidazole derivative that selectively and reversibly blocks small conductance Ca2+-activated K+ channels in a Ca2+-dependent manner.

Packaged Under Inert Gas

Specifications

Color White
Description 98% by HPLC
Molecular Formula C17H17N3
Formula Weight 263.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg