missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Normal Rabbit Serum, Lyophilized Solid

Catalog No. 8690195ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8690195ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 8690195ML Supplier MilliporeSigma™ Supplier No. 8690195ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suitable for use in radioimmunoassay. MilliporeSigma™; Calbiochem™ Normal Rabbit Serum, Lyophilized Solid is normal carrier sera, produced from pooled lots of sera from healthy, non-immunized rabbits.

  • Form: Lyophilized solid

Specifications

Content And Storage Ok to freeze. Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Species Rabbit
Form Lyophilized
Product Type Rabbit Serum
Quantity 5 mL
For Use With (Application) Radioimuunoassay
Hemoglobin Concentration <0.05%
Lyophilized Yes
pH 6.5 to 8