Learn More
MilliporeSigma™ Calbiochem™ Normal Rabbit IgG
Catalog No. NI01100UG
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
NI01100UG 100 μg
1 options
Catalog No. NI01100UG Supplier MilliporeSigma™ Supplier No. NI01100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Used in flow cytometry, immunoblotting, Immunofluorescence, immunohistochemistry, immunoprecipitation.
  • Derived from rabbit serum
  • Glycoprotein found in serum tissue fluids
  • Negative control in parallel with specific rabbit IgG-type primary antibodies
Specifications
Quantity 100 μg
Content And Storage 2°C to 8°C