missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Normal Mouse Serum

Catalog No. NS03L1ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NS03L1ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NS03L1ML Supplier MilliporeSigma™ Supplier No. NS03L1ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used in ELISA, Flow Cytometry, Immunoblotting, Immunofluorescence, Immunohistochemistry, Immunoprecipitation for the detection of Mouse Serum. MilliporeSigma™; Calbiochem™ Normal Mouse Serum is normal carrier sera from healthy, non-immunized mice.

  • Form: Lyophilized
  • Formulation: Lyophilized serum. Lyophilized from 250mM sodium chloride, 10mM sodium phosphate
  • Preservative: ≤0.1% sodium azide

Specifications

Content And Storage Ok to freeze. Reconstitute the lypohilized serum in 1 ml of sterile distilled water with occasional gentle mixing for at least 2 h at 4°C. Following reconstitution, aliquot and freeze (-20°C) for long-term storage or refrigerate (4°C) with 0.1% azide for short-term storage. Avoid freeze/thaw cycles of solutions.
Species Mouse
Form Lyophilized
Product Type Mouse Serum
Quantity 1 mL
Formulation Lyophilized serum. Lyophilized from 250mM sodium chloride, 10mM sodium phosphate
For Use With (Application) ELISA, Flow Cytometry, Immunoblotting, Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Lyophilized Yes
pH 7.6