missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Nocodazole
Description
Inhibitor of mitosis. Has highly specific antimicrotubular activity for mammalian cells in culture. Promotes tubulin depolymerization. Also available as a 50mm solution in DMSO (Cat. No. 487929).
Specifications
Specifications
| Color | Pale Yellow |
| Description | 99% by HPLC |
| Product Type | Inhibitor |
| Target Proteins | Promotes tubulin depolymerization |
| CAS | 31430-18-9 |
| Molecular Formula | C14H11N3O3S |
| Formula Weight | 301.3g/mol |
| Solubility | DMSO |
| Form | Solid |
| Quantity | 10 mg |