missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ NAE Inhibitor, MLN4924

Catalog No. 5054770001
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5054770001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5054770001 Supplier MilliporeSigma™ Supplier No. 5.05477.0001

NAE Inhibitor, MLN4924, CAS 951950-33-7, is an AMP mimetic that forms a NAE-catalyzed NEDD8 adduct, which acts as a tight-binding, ATP-competitive NAE inhibitor (IC50 = 4.7nM).

A cell-permeable AMP mimetic that targets the NEDD8-activating E1 enzyme NAE nucleotide-binding site and undergoes a NAE-catalyzed covalent NEDD8 adduct formation, the adduct in turn acts as a tight-binding, ATP-competitive NAE inhibitor (IC50 = 4.7nM), exhibiting much reduced or little potency against UAE/UBA1, UBA6/UBE1L2, SAE, ATG7, adenosine receptors A1/A2A/A2B/A3, or a panel of 12 cellular kinases. Selectively reduces cellular Ubc12-NEDD8, but not Ubc9-SUMO or Ubc10-Ub, thioester formation (ICmax = 90nM in HCT-116 cultures in 24 h), resulting in cullin-RING ligases substrates elevation. Shown to inhibit the growth of various cancer cells both in cultures (IC50 from 50nM to 1.03μM) in vitro and in murine xenograft models (30 to 60mg/kg via s.c.) in vivo via apoptosis induction.

Specifications

Color Light Beige
Description 98% by HPLC
Target Proteins NEDD8-Activating Enzyme
Molecular Formula C21H25N5O4S
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg