missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Myeloperoxidase Inhibitor-I
Description
Packaged Under Inert Gas
Specifications
Specifications
| Color | Beige |
| Description | 95% by HPLC |
| Product Type | Inhibitor |
| Target Proteins | Myeloperoxidase |
| CAS | 5351-17-7 |
| Molecular Formula | C7H9N3O |
| Formula Weight | 151.2g/mol |
| Solubility | DMSO or Dilute Acids |
| Content And Storage | Protect from light |
| Form | Solid |
| Show More |