missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ mTOR Inhibitor XI, Torin1

Catalog No. 47599110MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47599110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47599110MG Supplier MilliporeSigma™ Supplier No. 47599110MG
Only null left

mTOR Inhibitor XI, Torin1, CAS 1222998-36-8, is a cell-permeable, highly potent, ATP-competitive inhibitor of mTOR and DNA-PK (IC50 = 4.32 and 6.34 nM, respectively).

A cell-permeable pyridinonequinoline derivative that acts as a highly potent, ATP-competitive inhibibitor against mTOR and DNA-PK (IC50 = 4.32 and 6.34 nM, respectively), while inhibiting PI 3-K only at much higher concentrations (IC50 =171 to 533nM). Effectively inhibits mTORC1-mediated S6K1 phosphorylation in MEF (IC50 = 2nM) in vitro , as well as mTORC2-mediated Akt and mTORC1-dependent S6 phosphorylations in murine lung and liver in vivo (up to 6 h post single 20mg/kg i.p. dose). Despite its poor in vivo stability (T1/2 = 4.52 h in mice; 10mg/kg i.p.), Torin1 is reported to completely suppress U87MG-derived tumor expansion in mice when administered via daily i.p. at a high dosage of 200mg/kg. Also reported to effectively inhibit cellular mTORC1 functionalities known to be rapamycin-resistant (Cat. Nos. 553210, 553211, and 553212).

Specifications

Color Yellowish-white
Description 99% by HPLC
Target Proteins MTOR
Molecular Formula C35H28F3N5O2
Formula Weight 607.62g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg