missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ MPS1 Inhibitor, NMS-P715

Catalog No. 4759495MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4759495MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4759495MG Supplier MilliporeSigma™ Supplier No. 4759495MG

The MPS1 Inhibitor, NMS-P715 controls the biological activity of MPS1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas

Specifications

Color White
Description 98% by HPLC
Molecular Formula C35H39F3N8O3
Formula Weight 676.31g/mol
Content And Storage Protect from light
Form Powder
Quantity 5 mg