missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ MMP-12 Inhibitor, MMP408

Catalog No. 4442915MG
Change view
Click to view available options
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
4442915MG 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4442915MG Supplier MilliporeSigma™ Supplier No. 4442915MG

The MMP-12 Inhibitor, MMP408, also referenced under CAS 1258003-93-8, controls the biological activity of MMP-12. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Packaged Under Inert Gas

Specifications

Color Off-white
Description 94% by HPLC
CAS 1258003-93-8
Molecular Formula C19H20N2O7S
Formula Weight 420.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg