missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Metformin

Catalog No. 3172405GM
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3172405GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3172405GM Supplier MilliporeSigma™ Supplier No. 3172405GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An antihyperglycemic agent that lowers blood glucose levels without stimulating insulin secretion, promoting weight gain or causing hypoglycemia.

Packaged Under Inert Gas

Specifications

Color White
Description 97% by Titration
Target Proteins AMP-activated protein kinase (AMPK)
CAS 1115-70-4
Molecular Formula C4H11N5·HCl
Formula Weight 165.6g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Solid
Quantity 5 g