missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Melatonin

Catalog No. 4443001GM
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4443001GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4443001GM Supplier MilliporeSigma™ Supplier No. 4443001GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A hormone that has been postulated as the mediator of photo-induced anti-gonadotropic activity in photoperiodic mammals.

A hormone that has been postulated as the mediator of photo-induced anti-gonadotropic activity in photoperiodic mammals. Counteracts the apoptotic effects of etoposide in bone marrow cells. Melatonin receptors are coupled to a G-protein system. Exhibits antioxidant properties. Has neuroprotective effects against Aβ peptides. Inhibits rat cerebellar nitric oxide synthase (NOS). Also acts as a peroxynitrite scavenger.

Specifications

Color Off-white
Description 98% by HPLC
Product Type Inhibitor
Target Proteins rat NOS
CAS 73-31-4
Molecular Formula C13H16N2O2
Formula Weight 232.3g/mol
Solubility EtOH
Content And Storage Protect from light
Form Solid
Quantity 1 g
Show More Show Less