missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ MEK1/2 Inhibitor III

Catalog No. 4449665MG
Change view
Click to view available options
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
4449665MG 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4449665MG Supplier MilliporeSigma™ Supplier No. 4449665MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The MEK1/2 Inhibitor III, also referenced under CAS 391210-10-9, controls the biological activity of MEK1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas

Specifications

Color Off-white
Description 95% by HPLC
CAS 391210-10-9
Molecular Formula C16H14F3IN2O4
Formula Weight 482.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg