missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Calpain Inhibitor VI

Catalog No. 2087451MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2087451MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2087451MG Supplier MilliporeSigma™ Supplier No. 2087451MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Calpain Inhibitor VI, also referenced under CAS 190274-53-4, controls the biological activity of Calpain. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: 4-Fluorophenylsulfonyl-Val-Leu-CHO
  • Cell Permeable

Specifications

Color White
Product Type Protease Inhibitor
Target Proteins μ-calpain
CAS 190274-53-4
Molecular Formula C17H25FN2O4S
Formula Weight 372.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Lyophilized
Quantity 1 mg
Purity 97% by HPLC
Show More Show Less