missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Lactimidomycin

Catalog No. 5062910001
Change view
Click to view available options
0.2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
5062910001 0.2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5062910001 Supplier MilliporeSigma™ Supplier No. 5.06291.0001

Lactimidomycin, CAS 134869-15-1, inhibits cell migration, proliferation, & protein synthesis (IC50 = 37.8nM). Blocks the first translocation step and has no effect on tRNA binding.

  • Packaged Under Inert Gas
  • Deactivate with 5% Bleach

Specifications

Color White
Description 98% by HPLC
Target Proteins Ribosome
Molecular Formula C26H35NO6
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 0.2 mg