missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Lactate Dehydrogenase A Inhibitor, FX11

Catalog No. 42721810MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42721810MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42721810MG Supplier MilliporeSigma™ Supplier No. 42721810MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lactate Dehydrogenase A Inhibitor, FX11, CAS 213971-34-7, is a cell permeable, selective, reversible, NADH competitive inhibitor of lactate dehhydrogenase A (LDHA; Ki = 8μM for human liver LDHA).

A cell permeable, gossypol analog that acts as a selective, reversible, and NADH competitive inhibitor of lactate dehhydrogenase A (LDHA; Ki = 8μM for human liver LDHA). Does not affect the activities of LDHB and glyceraldehyde-3-phosphate dehydrogenase even at higher concentrations. Shown to increase oxygen consumption, reactive oxygen species production, and cell death in P493 human lymphoma B cells. Also shown to reduce mitochondrial transmembrane potential and ATP levels and increase AMP kinase activity. Exhibits synergistic toxicity in P493 cells when combined with FK866 (Cat. No. 481908). Blocks the progression of human lymphoma and pancreatic cancer xenografts and when used in combination with FK866 it induces lymphoma regression.

Specifications

Color White
Description 96% by HPLC
Target Proteins LDHA
Molecular Formula C22H22O4
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg