missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ L-(–)-Norepinephrine-(+)-bitartrate

Catalog No. 489350100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
489350100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 489350100MG Supplier MilliporeSigma™ Supplier No. 489350100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

α,β-Adrenergic agonist.

α,β-Adrenergic agonist. A vasoconstrictive agent that causes down-regulation of angiotensin II receptors. Oxidation sensitive.

Specifications

Color White
Description 99% by titration
Product Type Agonist
Target Proteins α,β-Adrenergic agonist
CAS 69815-49-2
Molecular Formula C8H11NO3·C4H6O6
Formula Weight 319.3g/mol
Solubility H2O
Content And Storage Protect from moisture
Form Crystalline Solid
Quantity 100 mg
Show More Show Less