missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ JAK3 Inhibitor VI

Catalog No. 4201265MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4201265MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4201265MG Supplier MilliporeSigma™ Supplier No. 4201265MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The JAK3 Inhibitor VI, also referenced under CAS 856436-16-3, controls the biological activity of JAK3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Color Yellow
Description 98% by HPLC
Product Type Inhibitor
Target Proteins JAK-3
CAS 856436-16-3
Molecular Formula C18H13N3O·MeSO3H
Formula Weight 383.4g/mol
Solubility DMSO or H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 5 mg
Show More Show Less