missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Y-27632

Catalog No. 688001500UG
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
688001500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 688001500UG Supplier MilliporeSigma™ Supplier No. 688001500UG

Y-27632A, CAS 146986-50-7, is a cell-permeable, reversible, inhibitor of Rho kinases (Ki = 140 nM for p160ROCK). Enhances survival & cloning efficiency of ESC without affecting their pluripotency.

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Formulation A 5 mM (500μg/296μl) solution of Y-27632 in H2O.
Description 95% by HPLC
Target Proteins p160ROCK
Molecular Formula C14H21N3O·2HCl·H2O
Formula Weight 338.3g/mol
Content And Storage Protect from light
Form Liquid
Quantity 500 μg