missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ TGF-β RI Kinase Inhibitor VI, SB431542

Catalog No. 6164645MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6164645MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6164645MG Supplier MilliporeSigma™ Supplier No. 6164645MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The InSolution™ TGF-β RI Kinase Inhibitor VI, SB431542 controls the biological activity of TGF-β RI. Primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas

Specifications

Formulation A 100 mM (5mg/119μl) solution of SB431542 in DMSO.
Description 97% by HPLC
CAS 301836-41-9
Molecular Formula C22H16N4O3·2H2O
Formula Weight 420.4g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 5 mg