missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Tamoxifen, 4-Hydroxy-, (Z)-

Catalog No. 5082250001
Change view
Click to view available options
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
5082250001 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5082250001 Supplier MilliporeSigma™ Supplier No. 5.08225.0001

A 10 mM (2 mg/516 μL) sterile-filtered solution of (Z)-4-Hydroxytamoxifen in ethanol.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach

Specifications

Formulation A 10 mM (2mg/516μL) sterile-filtered solution of (Z)-4-Hydroxytamoxifen in EtOH.
Description 99% by HPLC
Target Proteins Estrogen receptor
Molecular Formula C26H29NO2
Formula Weight 387.5g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 2 mg