missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Smoothened Agonist, SAG

Catalog No. 566661500UG
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
566661500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 566661500UG Supplier MilliporeSigma™ Supplier No. 566661500UG

InSolution™ Smoothened Agonist, SAG, is a 10 mM (500 μg/86 μl) solution of Smoothened Agonist, SAG (Cat. No. 566660) in H2O.

Packaged Under Inert Gas

Specifications

Formulation A 10 mM (500μg/86μl) solution of Smoothened Agonist, SAG in H2O.
Description 95% by HPLC
Molecular Formula C28H28ClN3OS·2HCl·H2O
Formula Weight 581g/mol
Content And Storage Protect from light
Form Liquid
Quantity 500 μg