missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Purmorphamine

Catalog No. 5402235MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5402235MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5402235MG Supplier MilliporeSigma™ Supplier No. 5402235MG

Purmorphamine is a cell-permeable purine compound that induces osteoblast differentiation in C3H10T1/2 multipotent mesenchymal progenitor cells.

Packaged Under Inert Gas

Specifications

Formulation A 50 mM (5mg/192μl) solution of Purmorphamine in DMSO.
Description 98% by HPLC
Molecular Formula C31H32N6O2
Formula Weight 520.6g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 5 mg