missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Nocodazole

Catalog No. 48792910MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48792910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48792910MG Supplier MilliporeSigma™ Supplier No. 48792910MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibitor of mitosis. Has highly specific antimicrotubular activity for mammalian cells in culture.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach

Specifications

Formulation A 50 mM (10mg/664μL) solution of Nocodazole in DMSO.
Description 97% by HPLC
Target Proteins Microtubules
CAS 31430-18-9
Formula Weight 301.3g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 10 mg