missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ MEK1/2 Inhibitor III, PD0325901

Catalog No. 4449685MG
Change view
Click to view available options
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
4449685MG 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4449685MG Supplier MilliporeSigma™ Supplier No. 4449685MG

The InSolution™ MEK1/2 Inhibitor III, PD0325901, also referenced under CAS 391210-10-9, controls the biological activity of MEK1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas

Specifications

Formulation A 50 mM (5mg/207μl) solution of MEK1/2 Inhibitor III, PD0325901 in DMSO.
Description 95% by HPLC
CAS 391210-10-9
Molecular Formula C16H14F3IN2O4
Formula Weight 482.2g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 5 mg