missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Jasplakinolide, Jaspis johnstoni

Catalog No. 42012750UG
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42012750UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42012750UG Supplier MilliporeSigma™ Supplier No. 42012750UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

InSolution™ Jasplakinolide, Jaspis johnstoni, CAS 102396-24-7, is a 1mm solution of Jasplakinolide, Jaspis johnstoni in DMSO. Acts as a potent inducer of actin polymerization and stabilization.

InSolution™ Jasplakinolide, Jaspis johnstoni, CAS 102396-24-7, is a 1mm solution of Jasplakinolide, Jaspis johnstoni in DMSO. Acts as a potent inducer of actin polymerization and stabilization.

Specifications

Formulation A 1 mM (50μg/71μl) solution of Jasplakinolide, Jaspis johnstoni in DMSO.
Description 90% by HPLC
Target Proteins F-actin
Molecular Formula C36H45BrN4O6
Formula Weight 709.7g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 50 μg