missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Cyclopamine-KAAD

Catalog No. 23980750ug
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23980750UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23980750UG Supplier MilliporeSigma™ Supplier No. 23980750UG

A cell-permeable potent analog of Cyclopamine that specifically inhibits Hedgehog (Hh) signaling with similar or lower toxicity.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach

Specifications

Formulation A 1 mM (50μg/72μL) solution of Cyclopamine-KAAD in DMSO.
Description 70% by HPLC (sum of two isomers)
Target Proteins SmoA1
Formula Weight 698g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 50 μg