missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Casein Kinase II Inhibitor I

Catalog No. 2187085MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2187085MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2187085MG Supplier MilliporeSigma™ Supplier No. 2187085MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The InSolution™ Casein Kinase II Inhibitor I controls the biological activity of Casein Kinase II. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Formulation A 10 mM (5mg/1.15mL) solution of Casein Kinase II Inhibitor I in DMSO.
Description Single spot by TLC
Product Type Inhibitor
Target Proteins CK2 rat liver, human recombinant
Molecular Formula C6HBr4N3
Formula Weight 434.7g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 5 mg