missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ InSolution™Blebbistatin, Racemic

Catalog No. 2033895MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2033895MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2033895MG Supplier MilliporeSigma™ Supplier No. 2033895MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

InSolution™Blebbistatin, Racemic, is a 50mm in 90% DMSO. A cell-permeable, selective, and reversible inhibitor of nonmuscle myosin II. Blocks cell blebbing and disrupts cell migration and cytoKinesis.

InSolution™Blebbistatin, Racemic, is a 50mm in 90% DMSO. A cell-permeable, selective, and reversible inhibitor of nonmuscle myosin II. Blocks cell blebbing and disrupts cell migration & cytoKinesis.

Specifications

Formulation A 50 mM (5mg/342μl) solution of (±)-Blebbistatin in 90% DMSO.
Description 97% by HPLC
Target Proteins ATPase
Molecular Formula C18H16N2O2
Formula Weight 292.3g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 5 mg