missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Genistein, Soybean

Catalog No. 34583420MG
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34583420MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34583420MG Supplier MilliporeSigma™ Supplier No. 34583420MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A cell-permeable, reversible, substrate competitive inhibitor of protein tyrosine kinases, including autophosphorylation of epidermal growth factor receptor kinase (IC50 = 2.6 μM).

Cell Permeable

Specifications

Color White
Product Type Inhibitor
Target Proteins EGFR kinase
CAS 446-72-0
Molecular Formula C15H10O5
Formula Weight 270.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 20 mg
Purity 95% by HPLC
Show More Show Less