missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ InSolution™ Aphidicolin
Description
- Packaged Under Inert Gas
- Cell Permeable
- Deactivate with 5% Bleach
Specifications
Specifications
| Formulation | A 30 mM (1mg/98μL) sterile-filtered solution of Aphidicolin in DMSO. |
| Description | 98% by TLC |
| Target Proteins | DNA polymerase α and δ |
| Molecular Formula | C20H34O4 |
| Formula Weight | 338.5g/mol |
| Content And Storage | Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light |
| Form | Liquid |
| Quantity | 1 mg |