missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ γ-Secretase Inhibitor X

Catalog No. 565771500UG
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
565771500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 565771500UG Supplier MilliporeSigma™ Supplier No. 565771500UG

InSolution™ γ-Secretase Inhibitor X is a 1 mM solution in DMSO. A highly potent inhibitor of g-secretase (IC50 = 17, 48, 67 nM for Aβtotal, Aβ40 & Aβ42 in SHSY5Y cells overexpressing spβA4CTF).

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Formulation A 1 mM (250μg in 372μl or 500μg in 744μl) solution in DMSO.
Target Proteins Aβtotal
Molecular Formula C39H52N4O6
Formula Weight 672.9g/mol
Content And Storage Protect from Light, Protect from Moisture
Form Liquid
Quantity 500 μg
Purity 90% by HPLC