missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ PPACK, Dihydrochloride
Description
- Packaged Under Inert Gas
- Peptide Sequence: H-D-Phe-Pro-Arg-chloromethylketone, 2HCl
Specifications
Specifications
| Color | White |
| Target Proteins | Thrombin |
| CAS | 142036-63-3 |
| Molecular Formula | C21H33N6O3Cl3 |
| Formula Weight | 523.9g/mol |
| Solubility | H2O |
| Content And Storage | Hygroscopic, Protect from Light, Protect from Moisture |
| Form | Solid |
| Quantity | 5 mg |
| Purity | 80% by HPLC |