missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ PPACK, Dihydrochloride

Catalog No. 5202225MG
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5202225MG 5 mg
52022225MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5202225MG Supplier MilliporeSigma™ Supplier No. 5202225MG

PPACK, Dihydrochloride, CAS 142036-63-3, is an extremely potent and selective irreversible inhibitor of thrombin (Kobs/[I] = 107M-1S-1). Reacts with thrombin in a 1:1 stoichiometry.

  • Packaged Under Inert Gas
  • Peptide Sequence: H-D-Phe-Pro-Arg-chloromethylketone, 2HCl

Specifications

Color White
Target Proteins Thrombin
CAS 142036-63-3
Molecular Formula C21H33N6O3Cl3
Formula Weight 523.9g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from Light, Protect from Moisture
Form Solid
Quantity 5 mg
Purity 80% by HPLC